Learn More
Medchemexpress LLC N4-acetylcytidine | 3768-18-1 | MFCD00006540 | 99.7% | 285.25 g/mol | C11H15N3O6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01967010MM/1ML
Encompass_Preferred
N4-acetylcytidine is a naturally occurring RNA nucleoside modification used in molecular biology and biochemical research to study RNA stability, translation, and metabolism. Offered as a ready-to-use 10 mM solution in DMSO (1 mL) and as a solid, the material is intended for research applications and analytical assays.
  • Ready-to-use 10 mM solution in DMSO for convenient dosing
  • Available as solid for custom formulation or storage
  • High purity for reproducible experimental results
  • Suitable for studies of RNA modification, stability, and translation
  • Documentation available, including COA and RP-HPLC data
Catalog No. 50-003-01848