Learn More
Apexbio Technology LLC N4-Acetylsulfamethoxazole 5mg
SDP
Supplier:  Apexbio Technology LLC BA15635
Encompass_Preferred
N4-Acetylsulfamethoxazole (CAS 21312-10-7) is a primary metabolite of sulfamethoxazole a sulfonamide antimicrobial agent This compound results from the N4-acetylation of sulfamethoxazole during hepatic metabolism and represents the predominant excreted form in urine While N4-Acetylsulfamethoxazole does not possess intrinsic antibacterial activity it is utilized in biomedical research to investigate the pharmacokinetics metabolism and disposition of sulfonamide drugs Studies involving this metabolite aid in understanding drug biotransformation pathways and can inform the evaluation of drug safety and renal excretion profiles
Catalog No. 50-000-91404