Learn More
Medchemexpress LLC N6-(1-carboxyethyl)-L-lysine | 5746-03-2 | MFCD30364298 | 99.9% | 218.25 | C9H18N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW58587410MG
Encompass_Preferred
Nε-(Carboxyethyl)lysine (CEL) is an advanced glycation end product used in biochemical and cellular research to study protein glycation, receptor for AGE (RAGE) signalling, and related pathological processes. It is supplied as a white to off-white solid with high analytical purity for laboratory applications.
  • High purity (99.9%) suitable for analytical and cellular studies.
  • Used to model advanced glycation end product formation and protein cross-linking.
  • Interacts with RAGE receptor pathways to study inflammatory signaling.
  • Modulates glutamate transporters and affects neurotransmission-related assays.
  • Characterized chemical identity with CAS 5746-03-2 and defined molecular weight.
  • Provided as a solid for ease of handling and storage.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-84286