Learn More
Medchemexpress LLC N6-((2-azidoethoxy)carbonyl)-L-lysine | 1167421-25-1 | MFCD22393077 | 98.0% | 259.26 g/mol | C9H17N5O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1114345MG
Encompass_Preferred
UAA crosslinker 1 (N6-((2-azidoethoxy)carbonyl)-L-lysine) is an azide-containing noncanonical amino acid used for site-specific incorporation into proteins and for bioorthogonal crosslinking. It enables conjugation through click chemistry reactions and is suitable for biochemical and bioconjugation applications.
Catalog No. 50-002-63395