Learn More
Medchemexpress LLC N6-(2-hydroxyethyl)adenosine | 4338-48-1 | 311.30 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00695710MM/1ML
Encompass_Preferred
N6-(2-Hydroxyethyl)adenosine is a purine nucleoside analog that inhibits the NF-κB/Smad signaling pathway. It demonstrates anti-hyperglycemia, antioxidant, antitumor, anti-inflammatory, and insecticidal activities and is orally active.
  • Modulates inflammatory responses, reducing pro-inflammatory cytokines and increasing anti-inflammatory ones.
  • Exhibits anti-fibrosis effects in kidney cells.
  • Induces cytotoxicity, apoptosis, and autophagy in various cancer cell lines.
  • Demonstrates insecticidal activity.
  • Ameliorates hyperglycemia and kidney damage in diabetic models.
  • Enhances antioxidant enzyme activities and reduces oxidative stress markers.
  • Shows antitumor activity by reducing tumor volume and weight in animal models.
Catalog No. 50-004-60327