Learn More
Medchemexpress LLC N6-(4-Hydroxybenzyl)adenosine | 110505-75-4 | 100.0% | 373.36 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1877510MM/1ML
Encompass_Preferred
N6-(4-Hydroxybenzyl)adenosine, also known as Para-topolin riboside, is an inhibitor of platelet aggregation induced in vitro by collagen. Its anti-aggregation activity is believed to involve an interaction with the P2Y12 receptor binding site, making it suitable for studies on platelet function and related pathways.
  • Inhibits platelet aggregation
  • IC50 range of 6.77-141 μM against collagen-induced aggregation
  • May interact with P2Y12 receptor
  • Prevents serum deprivation-induced PC12 cell apoptosis
  • Intended for research use only
Catalog No. 50-003-88223