Learn More
Medchemexpress LLC N6-(4-Hydroxybenzyl)adenosine | 110505-75-4 | 99.96% | 373.36 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY187755MG
Encompass_Preferred
N6-(4-Hydroxybenzyl)adenosine (Para-topolin riboside) is a chemical compound known for its inhibitory effects on platelet aggregation. It primarily acts on the P2Y12 receptor binding site and has shown activity against collagen-induced aggregation in vitro.
  • Inhibits platelet aggregation induced by collagen.
  • Interacts with the P2Y12 receptor binding site.
  • Prevents serum deprivation-induced PC12 cell apoptosis.
  • Inhibits collagen-induced human platelet aggregation.
Catalog No. 50-003-86097