Learn More
Medchemexpress LLC N6-(4-Hydroxybenzyl)adenosine | 110505-75-4 | 99.96% | 373.36 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1877550MG
Encompass_Preferred
N6-(4-Hydroxybenzyl)adenosine is an inhibitor of platelet aggregation induced in vitro by collagen. Its anti-aggregation activity may involve an interaction with the P2Y12 receptor binding site. It prevents serum deprivation-induced PC12 cell apoptosis.
  • Inhibits platelet aggregation
  • Prevents PC12 cell apoptosis
  • Target for P2Y12 receptor
  • Multiple solubility protocols available
  • Long-term storage stability for powder and solvent forms
Catalog No. 50-003-88226