Learn More
Medchemexpress LLC N6-(4-hydroxybenzyl)adenosine | 110505-75-4 | MFCD18641957 | 100.0% | 373.36 g/mol | C17H19N5O5 | 10 MG

Supplier: Medchemexpress LLC HY1877510MG
N6-(4-Hydroxybenzyl)adenosine is an adenosine derivative reported to inhibit platelet aggregation and potentially interact with P2Y12 receptor binding sites. It is supplied as a solid research-grade chemical with high purity for laboratory studies.
- Inhibits collagen-induced platelet aggregation in vitro.
- May interact with the P2Y12 receptor binding site.
- High purity: 99.96%.
- Molecular weight 373.36 g/mol; formula C17H19N5O5.
- Solid form; powder storage: -20°C for 3 years or 4°C for 2 years.
- Available in small laboratory pack sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.