Learn More
Cayman Chemical N6 4AmInobutyladenosIn5 2.5mg
SDP
Supplier:  Cayman Chemical 386362.5 MG
Encompass_Preferred
An aminobutyl derivative of ADP; has been used in the synthesis of quantum dots in ADP-specific TR-FRET assays
Catalog No. 50-291-6379