Learn More
Medchemexpress LLC N6-Benzoyladenosine | 4546-55-8 | 371.35 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW008388100G
Encompass_Preferred
N6-Benzoyladenosine (Bz-rA) is a nucleoside derivative with an N6 protecting group. It is primarily used as a synthetic intermediate to participate in photocatalytic oxidative cyclization reactions and synthesize oligoribonucleotides. N6-Benzoyladenosine can be used to efficiently construct heterocyclic skeletons in photochemical conversions and protect the ribose moiety to regulate the cyclization pathway.
  • Used as a synthetic intermediate in photocatalytic oxidative cyclization reactions
  • Synthesizes oligoribonucleotides
  • Constructs heterocyclic skeletons efficiently in photochemical conversions
  • Protects the ribose moiety to regulate the cyclization pathway
  • Has antiviral activity against Enterovirus 71 (EC50: 12 μM)
  • Reacts with 1 mM oxidant under irradiation with a 400 W high-pressure mercury lamp
Catalog No. 50-004-47194