Learn More
Medchemexpress LLC N6-benzoyladenosine | 4546-55-8 | 371.35 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00838850G
Encompass_Preferred
N6-Benzoyladenosine (Bz-rA) is a nucleoside derivative featuring an N6 protecting group. It primarily serves as a synthetic intermediate in photocatalytic oxidative cyclization reactions and in the synthesis of oligoribonucleotides. It is also utilized for efficient construction of heterocyclic skeletons in photochemical conversions and for protecting the ribose moiety to control the cyclization pathway.
  • Features N6 protecting group
  • Functions as a synthetic intermediate in photocatalytic oxidative cyclization reactions
  • Used for synthesizing oligoribonucleotides
  • Enables efficient construction of heterocyclic skeletons in photochemical conversions
  • Protects the ribose moiety to control the cyclization pathway
Catalog No. 50-004-47312