Learn More
Apexbio Technology LLC N6-Cyclopentyladenosine 41552-82-3 50mg
SDP
Supplier:  Apexbio Technology LLC B683850
Encompass_Preferred
N6-Cyclopentyladenosine is a small-molecule agonist targeting the adenosine A1 receptor It is designed to selectively activate the A1 receptor subtype thereby modulating intracellular signaling cascades involved in cardiovascular function neuroprotection and neurotransmission regulation N6-Cyclopentyladenosine exerts its biological activity primarily through activation of the adenosine A1 receptor Based on these pharmacological properties N6-Cyclopentyladenosine holds research potential in cardioprotection studies of ischemia-induced injury neurodegenerative conditions and neurotransmitter-related research
Catalog No. 50-000-89710