Learn More
Medchemexpress LLC HY-111809 50mg Medchemexpress, N6-Ethyladenosine CAS:14357-08-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11180950MG
Encompass_Preferred
Medchemexpress, HY-111809 50mg N6-Ethyladenosine CAS:14357-08-5 N6-Ethyladenosine is an adenosine derivative, acts as a Adenosine receptor agonist, with Kis of 4.9 and 4.7 nM for hA1AR and hA3AR, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8972