Learn More
Medchemexpress LLC N6-Methyl-dA phosphoramidite | 105931-58-6 | 99.8% | 767.85 | 50 MG

Supplier: Medchemexpress LLC HY13858250MG
N6-Methyl-dA phosphoramidite is a chemical reagent that can be used in the synthesis of oligodeoxyribonucleotides. It is intended for research use only.
- Appearance: Solid, white to off-white
- Molecular formula: C41H50N7O6P
- Used in DNA/RNA synthesis
- Related to oligonucleotides and phosphoramidites
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.