Learn More
Aviva Systems Biology Corp N6AMT1 Antibody - N-terminal region (ARP45845_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against N6AMT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human N6AMT1. Alternate names: C21orf127, HEMK2, MGC19995, MTQ2, N6AMT, PRED28ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59150