Learn More
Sino Biological Recombinant Human MLL3/KMT2C Protein (GST Tag), Active
Supplier:  Sino Biological M343381G500
Encompass_Preferred
Recombinant human MLL3 (4689-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4286