Learn More
Cayman Chemical N6benzoylCyclIc AMPsodIM 10mg
SDP
Supplier:  Cayman Chemical 1881910 MG
Encompass_Preferred
A cell permeable cAMP analog that selectively activates PKA over Epac1 and 2 (also known as RAPGEF3 and 4); binds the AI and AII subunits of PKA with pKi values of 4.0 and 3.8, respectively, whereas it binds Epac1 with a pKi value of 1.3
Catalog No. 50-296-6185