Learn More
Medchemexpress LLC NA-014 | 3041127-59-4 | 99.5% | 380.46 | 25 MG

Supplier: Medchemexpress LLC 25MGHY162455
NA-014 is a selective EAAT2 positive allosteric modulator (PAM), exhibiting an EC50 of 3 nM. It is categorized as an Inhibitor within the Membrane Transporter/Ion Channel pathway. This product is suitable for research use only and has not been fully validated for medical applications.
- Selective EAAT2 positive allosteric modulator (PAM)
- Target: EAAT
- Pathway: Membrane Transporter/Ion Channel
- EC50 of 3 nM
- Solid, light yellow to brown appearance
- Shipping at room temperature
- Store solid at 4°C under nitrogen
- Store in solvent at -80°C for 6 months or -20°C for 1 month under nitrogen
- Solubility in DMSO: 100 mg/mL (262.84 mM; requires ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.