Learn More
Cayman Chemical Na-Benzoyl-DL-ArgInIn-pNAh 1g
SDP
Supplier:  Cayman Chemical 339341 G
Encompass_Preferred
A colorimetric serine protease substrate; release of pNA can be quantified by colorimetric detection at 405 nm as a measure of serine protease activity
Catalog No. 50-295-0502