Learn More
Cayman Chemical Na-Boc-D-HIstIdIn 1g
SDP
Supplier:  Cayman Chemical 305011 G
Encompass_Preferred
An amino acid building block; has been used in the synthesis of antimicrobial peptides, thyrotropin-releasing hormone enantiomers, and proteolysis-resistant peptide renin inhibitors
Catalog No. 50-299-1659