Learn More
Cayman Chemical Na-Boc-D-HIstIdIn 500mg
SDP
Supplier:  Cayman Chemical 30501500 MG
Encompass_Preferred
An amino acid building block; has been used in the synthesis of antimicrobial peptides, thyrotropin-releasing hormone enantiomers, and proteolysis-resistant peptide renin inhibitors
Catalog No. 50-299-0640