Learn More
Strem, An Ascensus Company CAS# 4984-82-1. 0.5mole. Sodium cyclopentadienide, 2-3M in THF. MFCD00012161
SDP
Supplier:  Strem, An Ascensus Company 931075500MMOL
Encompass
CAS# 4984-82-1. 0.5mole. Sodium cyclopentadienide, 2-3M in THF. MFCD00012161. Molecular Weight: 88.09. Molecular Formula: C5H5Na. Color/form: yellow to red solution Strem# 93-1075. http://www.strem.com/catalog/v/93-1075/
Catalog No. 50-201-6793