Learn More
American Radiolabeled Chemicals Inc NA OXALATE 14C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 345850UC
Encompass_Preferred
Sodium oxalate-14C 50uCi in sterile water, 0.1 mCi/ml, S.A. 10-20 mCi/mmol, M.W. 134.10, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-268