Learn More
Aviva Systems Biology Corp Naa60 Antibody - N-terminal region (ARP48550_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48550_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Naa60. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Rat Naa60. Alternate names: MGC124897, RGD1308915ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-366