Learn More
Cayman Chemical NAADPsodIum salt 10mg
SDP
Supplier:  Cayman Chemical 2599210 MG
Encompass_Preferred
A secondary messenger that induces calcium mobilization; induces calcium release from endosomes and lysosomes via TPC2 and TPC1; induces calcium mobilization in sea urchin and starfish eggs post fertilization to block polyspermy and activate embryogenesis; induces VEGF-mediated angiogenesis in HUVECs; alkalizes lysosomal pH, arresting autophagic flux in mouse embryonic stem cells
Catalog No. 50-293-2818