Learn More
Selleck Chemical LLC Nabumetone S4051-1g
SDP
Supplier:  Selleck Chemical LLC S40511G
Encompass_Preferred
Nabumetone (BRL-14777) is a non-steroidal anti-inflammatory drug and its active metabolite inhibits the COX
Catalog No. 50-310-9650