Learn More
Apexbio Technology LLC Nabumetone 42924-53-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B14505.1
Encompass_Preferred
Nabumetone (CAS 42924-53-8) is a non-steroidal anti-inflammatory compound that undergoes hepatic metabolism to yield its active form 6-methoxy-2-naphthylacetic acid (6-MNA) The active metabolite selectively inhibits cyclooxygenase (COX) particularly COX-2 resulting in reduced synthesis of prostaglandins involved in inflammation and pain signaling Nabumetone is commonly utilized in research to study prostaglandin-mediated inflammatory pathways COX enzyme function and the pharmacological modulation of inflammatory responses in preclinical models
Catalog No. 50-000-91635