Learn More
Cayman Chemical nacardIc AcId 10mg
SDP
Supplier:  Cayman Chemical 1314410 MG
Encompass_Preferred
An alkyl salicylic acid isolated from cashew shells; inhibits the HAT activity of p300 and PCAF (IC50 = 8.5 and 5 µM, respectively); suppresses NF-κB activation, inhibits IκBα phosphorylation, and prohibits p65 nuclear translocation
Catalog No. 50-295-2436