Learn More
Cayman Chemical NacetylSgernylgernylLC 10mg
SDP
Supplier:  Cayman Chemical 6334010 MG
Encompass_Preferred
AGGC is a synthetic substrate for the isoprenylated protein methyltransferase (also known as S-adenosylmethionine-dependent methyltransferase).{2306,2304} Because it is able to serve as a substrate for the methyltransferase, it effectively functions as an inhibitor of methylation of endogenous isoprenylated proteins.
Catalog No. 50-289-4669