Learn More
YSI 3P LifeScience 2392 NaCl Solution (30 mL)
Supplier:  YSI 3P LifeScience 2392
Encompass
Sodium chloride solution. Use when installing membranes on YSI 2300, 2700, and 2900 Series Analyzers.
Catalog No. 50-200-154