Learn More
Medchemexpress LLC HY-B0445 500mg , NAD+ CAS:53-84-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0445500MG
Encompass_Preferred
HY-B0445 500mg NAD+ CAS: 53-84-9 NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9810