Learn More
Medchemexpress LLC NAD-dependent Glucose dehydrogenase (GDH-NAD) | 9028-53-9 | 1 KU
SDP
Catalog No. 5000399711
Encompass_Preferred
NAD-dependent Glucose dehydrogenase (GDH-NAD) (EC 1.1.1.47) is an NAD-dependent oxidoreductase. Glucose dehydrogenase (GDH) catalyzes the oxidation of β-D-glucose to β-D-glucono-1,5-lactone with simultaneous reduction of the cofactor NADP+ to NADPH or, to a lesser extent, NAD+ to NADH. This enzyme is versatile, accepting both NAD+ and NADP+ as cofactors, making it suitable for the regeneration of NADH and NADPH. It appears as a solid with a color ranging from white to off-white and is intended for research use only.
  • Catalyzes oxidation of β-D-glucose to β-D-glucono-1,5-lactone
  • Reduces NADP+ to NADPH or NAD+ to NADH
  • Accepts both NAD+ and NADP+ as cofactors
  • Suitable for regeneration of NADH and NADPH
  • Appears as a solid with a color ranging from white to off-white
  • Intended for research use only
Catalog No. 50-003-99711 Supplier Medchemexpress LLC Supplier No. HYP3186A1KU
NAD-dependent Glucose dehydrogenase (GDH-NAD) (EC 1.1.1.47) is an NAD-dependent oxidoreductase. Glucose dehydrogenase (GDH) catalyzes the oxidation of β-D-glucose to β-D-glucono-1,5-lactone with simultaneous reduction of the cofactor NADP+ to NADPH or, to a lesser extent, NAD+ to NADH. This enzyme is versatile, accepting both NAD+ and NADP+ as cofactors, making it suitable for the regeneration of NADH and NADPH. It appears as a solid with a color ranging from white to off-white and is intended for research use only.
  • Catalyzes oxidation of β-D-glucose to β-D-glucono-1,5-lactone
  • Reduces NADP+ to NADPH or NAD+ to NADH
  • Accepts both NAD+ and NADP+ as cofactors
  • Suitable for regeneration of NADH and NADPH
  • Appears as a solid with a color ranging from white to off-white
  • Intended for research use only