Learn More
Sigma Aldrich Fine Chemicals Biosciences NAD Grade II free acid
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 10621650001
Encompass
Nicotinamide adenine dinucleotide (NAD) is a cofactor containing an adenylic acid and a nicotinamide-5AE-ribonucleotide group linked together by a pyrophosphate moiety. It is ubiquitously found. In NAD complexes the enzyme-cofactor interactions are highly conserved.
Catalog No. 50-290-3855