Learn More
Gold Biotechnology Inc NADH Beta
SDP
Supplier:  Gold Biotechnology Inc N0301
Encompass_Preferred
Nicotinamide adenine dinucleotide (NAD ) plays a major role in metabolism as a cofactor and mobile electron acceptor NAD is a required oxidizing cosubstrate for many enzymes It is reduced to which carries electrons to the electron transport chain for subsequent oxidative phorphorylation and ATP production NAD is capable of donating ADP-ribose moieties to a protein producing nicotinamide in the process Sirtuin enzymes use NAD as a substrate to deacetylate proteins and direct activity between the nucleus and mitochondria NAD is regenerated by fermentation and by oxidative phosphorylation
Catalog No. 50-005-62100