Learn More
Medchemexpress LLC Nadifloxacin | 124858-35-1 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB050610MM/1ML
Encompass_Preferred
Nadifloxacin is a broad-spectrum quinolone antibiotic that inhibits bacterial DNA gyrase and topoisomerase IV, thereby interfering with DNA replication. It also suppresses the production of proinflammatory cytokines such as IL-1α, IL-6, and IL-8. This compound demonstrates antibacterial activity against various pathogens, including *Propionibacterium acnes* and *Staphylococcus aureus*, and also exhibits anti-inflammatory activity. It is utilized in the research of skin infections like acne vulgaris, folliculitis, and impetigo.
  • Inhibits bacterial DNA gyrase and topoisomerase IV
  • Suppresses proinflammatory cytokines (IL-1α, IL-6, IL-8)
  • Exhibits antibacterial activity against *Propionibacterium acnes* and *Staphylococcus aureus*
  • Shows anti-inflammatory activity
  • Used in research for skin infections
  • Potent activity against *Propionibacterium acnes* isolated from acne vulgaris
Catalog No. 50-004-06840