Learn More
Medchemexpress LLC Nadolol | 42200-33-9 | 99.98% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB080410MM/1ML
Encompass_Preferred
Nadolol (SQ-11725) is a non-selective and orally active β-adrenergic receptor blocker. It is also a substrate of organic anion transporting polypeptide 1A2 (OATP1A2). This compound is valuable for research related to high blood pressure, angina pectoris, and vascular headaches. It is presented as a white to off-white solid.
  • Non-selective and orally active beta-adrenergic receptor blocker
  • Substrate of organic anion transporting polypeptide 1A2 (OATP1A2)
  • Suitable for research in hypertension, angina pectoris, and vascular headaches
  • Appearance: white to off-white solid
  • 1H NMR spectrum consistent with its structure
  • Complies with specified quality standards
Catalog No. 50-004-08084