Learn More
Research Products International Corp NADP [B-Nicotinamide Adenine Dinucleotide Phosphate, Oxidized Form, Monosodium Salt Trihydrate], 1 Gram

Supplier: Research Products International Corp N201201.0
- Substance Name: β-Nicotinamide Adenine Dinucleotide Phosphate, Oxidized Form, Monosodium Salt Trihydrate
- CAS Number: 1184-16-3
- Molecular Formula: C21H27N7O17P3Na • 2H2O
- Molecular Weight: 801.4 g/mol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.