Learn More
Medchemexpress LLC NADP sodium hydrate | 698999-85-8 | 98.8% | 783.40 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY113325A500MG
Encompass_Preferred
NADP sodium hydrate is the sodium salt hydrate form of NADP. NADP is a coenzyme involved in cellular electron transfer reactions in biological metabolism, alternately oxidized (NADP+) and reduced (NADPH), and can maintain cellular redox homeostasis and regulate many biological events, including cellular metabolism. NADPH is a universal electron donor that provides reducing ability for synthetic metabolic reactions and redox balance. NADPH plays a multifunctional role in regulating inflammation, redox homeostasis, and synthetic metabolism processes.
  • It is the sodium salt hydrate form of NADP.
  • A coenzyme in cellular electron transfer reactions.
  • Maintains cellular redox homeostasis.
  • Regulates biological events, including cellular metabolism.
  • Provides reducing ability for synthetic metabolic reactions.
  • Contributes to redox balance.
  • Regulates inflammation, redox homeostasis, and synthetic metabolism.
Catalog No. 50-004-37596