Learn More
Medchemexpress LLC NADPH Oxidase 4 Antibody (YA4513) | 67 kDa | 100 UL

Supplier: Medchemexpress LLC HYP84816100UL
NADPH Oxidase 4 Antibody (YA4513) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting to NADPH Oxidase 4.
- Mouse host.
- Human reactivity.
- Monoclonal antibody.
- IgG1 type.
- Affinity purified.
- Supplied in PBS with 0.05% sodium azide.
- Liquid appearance.
- Store at -20°C for 1 year; avoid repeated freeze/thaw cycles.
- Applications include IHC-P (1:200-1:1000), ICC/IF (1:200-1:1000), FC (1:200-1:400), and ELISA (1:10000).
- Predicted and observed molecular weight: 67 kDa.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.