Learn More
Medchemexpress LLC NADPH Oxidase 4 Antibody (YA4640) | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP80762A10UL
Encompass_Preferred
NADPH Oxidase 4 Antibody (YA4640) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to NADPH Oxidase 4. This antibody is affinity purified and supplied in a formulation of 10mM phosphate buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets NADPH Oxidase 4
  • Affinity purified
  • Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
Catalog No. 50-004-42872