Learn More
MOLTOX Molecular Toxicology Inc NADPH REGENSYS A, 15 mL size refers to final volume of S9 mix per pkg, G-6-P MgCl2-KCL in 0.1M Phosphate Buffer, pH 7.4, Store in Refrigerator
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 60200.15
Encompass
Convenient preformulated filter sterilized and prepackaged S9 mix components for preparation of 4 - 10% S9 mix. Regensys™ """"A"""" is formulated as described by Maron & Ames, Mut. Res. 113: 173, 1983. Size refers to final volume of S9 mix. Regensys™ """"B"""" consists of accurately weighed aliquots of crystalline NADP to add to corresponding Regensys™ """"A"""" for completion of the Regensys™ system. MOLTOX S9 preparations and cofactor reagents for use in metabolic activation studies are manufactured to rigorous standards of quality and performance and are used by leading industrial, government and academic laboratories worldwide.
Catalog No. 50-948-929