Learn More
Medchemexpress LLC NAE-IN-M22 100mg | 864420-54-2 | 363.32 g·mol^-1 | C20H24Cl2N2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY115537100MG
Encompass_Preferred
NAE-IN-M22 (CAS 864420-54-2) is a research-grade small molecule inhibitor of the NEDD8-activating enzyme (NAE). It is described as a potent, selective, and reversible inhibitor with micromolar cellular activity, suitable for biochemical and cell-based studies.
  • Potent and selective NAE inhibitor.
  • Reversible mechanism with micromolar activity in cell assays.
  • Demonstrated induction of apoptosis in cancer cell lines, including A549.
  • Available in solid form and as a 10 mM solution in DMSO for flexible use.
  • High stated purity (~99.9%), appropriate for research applications.
Catalog No. 50-002-27127