Learn More
Medchemexpress LLC Nafamostat formate salt-13C6 | 81525-10-2 | 98.11% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYB0190S1500UG
Encompass_Preferred
Nafamostat formate salt-13C6 is the 13C labeled Nafamostat. Nafamostat is a synthetic serine protease inhibitor and an anticoagulant. It suppresses T cell auto-reactivity by decreasing granzyme activity and CTL cytolysis, and also blocks the activation of SARS-CoV-2.
  • 13C labeled Nafamostat
  • Synthetic serine protease inhibitor
  • Anticoagulant
  • Suppresses T cell auto-reactivity
  • Decreases granzyme activity and CTL cytolysis
  • Blocks activation of SARS-CoV-2
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07418