Learn More
Selleck Chemical LLC Nafamostat Mesylate 100mg 82956-11-4 FUT-175
SDP
Supplier:  Selleck Chemical LLC S1386100MG
Encompass_Preferred
Nafamostat mesilate (FUT-175) is a synthetic serine protease inhibitor, used as an anticoagulant during hemodialysis. Nafamostat mesylate blocks activation of SARS-CoV-2 and is investigated as a new treatment option for COVID-19. Nafamostat Mesilate attenuates inflammation and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1199