Learn More
Enzo Life Sciences Nafamostat . mesylate (10mg). CAS: 82956-11-4
SDP
Supplier:  Enzo Life Sciences BMLPI1430010
Encompass_Preferred
Competitive, reversible broad-spectrum serine proteinase inhibitor that can be a used as a specific tryptase inhibitor at very low concentrations. Has been used in vitro and in vivo. Purity: ≥98% (HPLC). Solubility: Soluble in water (25 mg/ml) Note: Phosphate-containing buffers may cause precipitation. Long Term Storage: Ambient.
Catalog No. 50-201-1482