Learn More
Abcam Nafamostat Mesylate, Protease inhibitor, 10MG
Supplier:  Abcam AB14143210MG
Encompass_Preferred
MW 539.6 Da, Purity >97%. Broad spectrum, potent protease inhibitor (Ki = 95.3 pM). Inhibits NF-κB activation and complement activation. Selective for human tryptase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6622