Learn More
Selleck Chemical LLC Nafcillin Sodium S4042-50mg
SDP
Supplier:  Selleck Chemical LLC S404250MG
Encompass_Preferred
Nafcillin sodium (CL 8491) reversibly inhibits -lactamase with Kd of 33 mM
Catalog No. 50-310-9551