Learn More
Apexbio Technology LLC Nafcillin Sodium 7177-50-6 10mM (in 1mL DMSO)
Supplier: Apexbio Technology LLC B19835.1
Nafcillin Sodium (7177-50-6) is a small-molecule inhibitor targeting -lactamase It is designed to reversibly inhibit -lactamase thereby preventing bacterial resistance mediated by hydrolysis of -lactam antibiotics Nafcillin Sodium exerts its biological activity primarily through reversible inhibition of -lactamase In in vitro studies Nafcillin Sodium demonstrates inhibitory activity with an IC50 value of approximately 33 mM Based on these pharmacological properties Nafcillin Sodium holds research potential in investigating -lactamase-mediated antibiotic resistance mechanisms evaluating therapeutic combination strategies and studying bacterial susceptibility profiles