Learn More
Medchemexpress LLC Nafcillin sodium monohydrate | 7177-50-6 | 98.1% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0555A5G
Encompass_Preferred
Nafcillin sodium monohydrate is an antibiotic and a reversible inhibitor of β-lactamase. It can be used for the research of staphylococcal infections.
  • Antibiotic
  • Reversible inhibitor of β-lactamase
  • Used for research of staphylococcal infections
  • Exhibits bactericidal activity against methicillin-susceptible Staphylococcus aureus (MSSA) and methicillin-resistant S. aureus (MRSA)
Catalog No. 50-004-09458